Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 2226 | 93 | |
XM_017001691.1 | XP_016857180.1 | 3 | 2131 | 93 | ||
GenBank mRNA | AF374386.1 | - | 3 | 2143 | 92 | |
AK000319.1 | - | 2 | 1947 | 93 | ||
AK024774.1 | - | 1 | 1646 | 93 | ||
BC001959.1 | - | 3 | 2193 | 3536977 | 93 | |
BC078177.1 | - | 3 | 2173 | 6526331 | 93 | |
BC085018.1 | - | 3 | 2196 | 6650445 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00092815 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 367864 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 98627 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 210527 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 182003 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3323742 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P103201 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2678 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P103201 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2679 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 671 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28859 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12537 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P103201 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3323742 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00801535 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3323742 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P103201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15038840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00092816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00092815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15038840 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00092815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00092816 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P103201 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00092816 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P103201 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 1593101 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 1593101 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P58190 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8039934 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898911 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8039935 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898912 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898914 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898915 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7898913 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8039936 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8039933 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7898910 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 217779_PM_s_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 222406_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11758304_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11716988_s_at | - | - |
Affymetrix Human Genome U219 Array | 11758304_s_at | - | - |
Affymetrix Human Genome U219 Array | 11716988_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11716989_s_at | - | - |
Affymetrix Human Genome U219 Array | 11716989_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01004621.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004621.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004621.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004621.hg.1 | ENST00000471915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004621.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004619.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004619.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004619.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004619.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004629.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004629.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004629.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004624.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004624.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004624.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004624.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004626.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004626.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004626.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01004634.hg.1 | TC01000305.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01004634.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004634.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004634.hg.1 | ENST00000471915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01004634.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151126.hg.1 | TC0100007344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151133.hg.1 | TC0100007344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000471915.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000579103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000471915.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | ENST00000579103.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151129.hg.1 | TC0100007344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151135.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151135.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151135.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151135.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151135.hg.1 | TC0100007344.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000471915.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000334351.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000374468.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | ENST00000471915.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | NM_017761.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151128.hg.1 | TC0100007344.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 217779_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 222406_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 217779_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 222406_s_at | - | - |
Human Genome U133A 2.0 Array | 217779_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 217779_s_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_017761_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw569739 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 217779_s_at10 | - | - |
Human Genome U95 Set | 65944_PM_at | - | - |
Human Genome U95 Set | 65945_PM_at | - | - |
Human Genome U95 Set | 65944_at | - | - |
Human Genome U95 Set | 65945_at | - | - |
Human Genome U133 Set | 217779_PM_s_at | - | - |
Human Genome U133 Set | 217779_s_at | - | - |
Human Genome U133 Set | 222406_s_at | - | - |
Human Genome U133 Set | 222406_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017927 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017927 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 192720 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2147306 | BC078177.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2147306 | BC085018.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2147306 | BC001959.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2147306 | BC068475.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2147306 | AK024774.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778957 | BC001959.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20127576-S | BC078177.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20127576-S | BC085018.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20127576-S | BC001959.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20127576-S | BC068475.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_20127576-S | AK024774.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_7879-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0002640019 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0002640019 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1500519 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_7879 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2147306 | AK024774.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778957 | BC001959.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1778957 | AF151042.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147306 | BC078177.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147306 | BC085018.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147306 | BC001959.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147306 | BC068475.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2147306 | AK024774.1 | human_transcriptome |