Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014966.3 | NP_055781.2 | 6 | 653 | 95 | |
NM_138615.2 | NP_619520.1 | 5 | 624 | 95 | ||
XM_006713033.1 | XP_006713096.1 | 4 | 447 | 95 | ||
XM_011533490.1 | XP_011531792.1 | 7 | 879 | 95 | ||
XM_011533491.1 | XP_011531793.1 | 8 | 1066 | 95 | ||
XM_011533492.2 | XP_011531794.1 | 6 | 898 | 95 | ||
XM_011533493.1 | XP_011531795.1 | 7 | 879 | 95 | ||
XM_011533494.2 | XP_011531796.1 | 5 | 588 | 95 | ||
XM_011533495.1 | XP_011531797.1 | 5 | 433 | 95 | ||
XM_011533496.1 | XP_011531798.1 | 6 | 785 | 95 | ||
XM_011533497.2 | XP_011531799.1 | 6 | 666 | 95 | ||
XM_011533498.1 | XP_011531800.1 | 6 | 736 | 95 | ||
XM_017005914.1 | XP_016861403.1 | 6 | 529 | 95 | ||
XM_017005915.1 | XP_016861404.1 | 4 | 404 | 95 | ||
XM_017005916.1 | XP_016861405.1 | 5 | 600 | 95 | ||
XM_017005917.1 | XP_016861406.1 | 7 | 695 | 95 | ||
GenBank mRNA | AB020697.1 | - | 5 | 341 | 95 | |
AK002076.1 | - | 2 | 336 | 95 | ||
AK096791.1 | - | 6 | 424 | 95 | ||
AK291266.1 | - | 6 | 428 | 95 | ||
BC015029.1 | - | 6 | 399 | 3911049 | 95 | |
BC020126.1 | - | 5 | 372 | 4339419 | 95 | |
BC038417.1 | - | 6 | 400 | 5171702 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |