Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008902.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008902.hg.1 |
ENST00000448920.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008902.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008902.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008902.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008909.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008909.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008909.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008909.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008909.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
ENST00000536088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008913.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
ENST00000536088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008916.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008910.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008910.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008910.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008910.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008910.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008908.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008908.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008908.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008908.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
TC0X000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
ENST00000483584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
ENST00000536088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
ENST00000544766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008912.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158472.hg.1 |
TC0X00008611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158470.hg.1 |
TC0X00008611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158478.hg.1 |
TC0X00008611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000298296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000409007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000443323.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_138702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008611.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
ENST00000483584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
ENST00000544766.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
NM_177456.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158467.hg.1 |
TC0X00008611.hg.1 |
Clariom D TC |