Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Hs02576842_m1
See other SORBS2 GE Assays ›
Gene Symbol:
SORBS2
Gene Name:
sorbin and SH3 domain containing 2
Gene Aliases:
ARGBP2, PRO0618
Chromosome Location:
Chr.4: 185585444 - 185956716 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs02576842_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SORBS2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001145670.1
NP_001139142.1
20 - 21
2681
122
NM_001145671.2
NP_001139143.1
22 - 23
2470
122
NM_001145672.1
NP_001139144.1
20 - 21
2108
122
NM_001145673.1
NP_001139145.1
22 - 23
2994
122
NM_001145674.1
NP_001139146.1
14 - 15
3281
122
NM_001145675.1
NP_001139147.1
14 - 15
1745
122
NM_001270771.1
NP_001257700.1
23 - 24
4316
122
NM_003603.6
NP_003594.3
22 - 23
2709
122
NM_021069.4
NP_066547.1
20 - 21
4096
122
XM_005263307.1
XP_005263364.1
24 - 25
3588
122
XM_005263308.1
XP_005263365.1
23 - 24
2946
122
XM_005263310.1
XP_005263367.1
23 - 24
2802
122
XM_005263311.1
XP_005263368.1
22 - 23
2787
122
XM_005263312.1
XP_005263369.1
22 - 23
2784
122
XM_005263313.1
XP_005263370.1
20 - 21
2685
122
XM_006714388.1
XP_006714451.1
23 - 24
2877
122
XM_006714390.1
XP_006714453.1
21 - 22
2694
122
XM_017008741.1
XP_016864230.1
27 - 28
4911
122
XM_017008748.1
XP_016864237.1
24 - 25
4611
122
XM_017008750.1
XP_016864239.1
26 - 27
4146
122
XM_017008751.1
XP_016864240.1
25 - 26
4062
122
XM_017008752.1
XP_016864241.1
25 - 26
3987
122
XM_017008753.1
XP_016864242.1
23 - 24
3846
122
XM_017008754.1
XP_016864243.1
22 - 23
3801
122
XM_017008755.1
XP_016864244.1
21 - 22
3576
122
XM_017008756.1
XP_016864245.1
20 - 21
3279
122
XM_017008757.1
XP_016864246.1
16 - 17
2681
122
XM_017008758.1
XP_016864247.1
23 - 24
2980
122
XM_017008759.1
XP_016864248.1
20 - 21
3407
122
XM_017008760.1
XP_016864249.1
22 - 23
2853
122
XM_017008762.1
XP_016864251.1
21 - 22
2730
122
XM_017008763.1
XP_016864252.1
20 - 21
2680
122
XM_017008764.1
XP_016864253.1
21 - 22
2661
122
XM_017008765.1
XP_016864254.1
22 - 23
2964
122
XM_017008766.1
XP_016864255.1
20 - 21
2637
122
XM_017008767.1
XP_016864256.1
21 - 22
2940
122
XM_017008768.1
XP_016864257.1
20 - 21
2858
122
XM_017008769.1
XP_016864258.1
20 - 21
2616
122
XM_017008770.1
XP_016864259.1
19 - 20
2592
122
XM_017008771.1
XP_016864260.1
21 - 22
2734
122
XM_017008772.1
XP_016864261.1
20 - 21
2568
122
XM_017008773.1
XP_016864262.1
21 - 22
2871
122
XM_017008774.1
XP_016864263.1
16 - 17
2361
122
XM_017008775.1
XP_016864264.1
25 - 26
4101
122
GenBank mRNA
AB018320.1
-
20 - 21
3748
122
AF049884.1
-
22 - 23
2673
122
AK056628.1
-
23 - 24
3934
122
AK225327.1
-
20 - 21
2647
122
AK225812.1
-
22 - 23
2994
122
AK293400.1
-
20 - 21
2108
122
AK296461.1
-
14 - 15
3058
122
AK304653.1
-
14 - 15
1481
122
AU142147.1
-
2 - 3
189
122
BC011883.1
-
22 - 23
2437
3677070
122
DA139168.1
-
1 - 2
84
122
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
3' Most
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene SORBS2
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
cell adhesion
biological_process
cell migration
cell growth involved in cardiac muscle cell development
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
structural constituent of muscle
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0