Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002041.hg.1 |
TC0X000177.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002041.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002041.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002041.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002043.hg.1 |
TC0X000177.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002043.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002043.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002042.hg.1 |
TC0X000177.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002042.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002042.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002042.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147489.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147489.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147489.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147489.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147489.hg.1 |
TC0X00006970.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147488.hg.1 |
TC0X00006970.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |