Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005510.hg.1 |
TC01000355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005510.hg.1 |
ENST00000321265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005510.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152414.hg.1 |
TC0100007489.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000321265.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000435827.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000452707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
ENST00000484772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007489.hg.1 |
NM_006600.3 |
Refseq |