Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007598.hg.1 |
ENST00000314634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007598.hg.1 |
ENST00000623907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007598.hg.1 |
ENST00000624571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007598.hg.1 |
NM_001004701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005489.hg.1 |
ENST00000314634.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005489.hg.1 |
NM_001004701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005489.hg.1 |
TC11000432.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100007598.hg.1 |
ENST00000314634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007598.hg.1 |
ENST00000623907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007598.hg.1 |
ENST00000624571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007598.hg.1 |
NM_001004701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000314634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000623907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000624571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
NM_001004701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000314634.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000623907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
ENST00000624571.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
NM_001004701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153766.hg.1 |
TC1100007598.hg.1 |
Clariom D TC |