Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015233.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015233.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015233.hg.1 |
ENST00000553795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015233.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015233.hg.1 |
TC10001222.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015236.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015236.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015236.hg.1 |
ENST00000553795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015236.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015236.hg.1 |
TC10001222.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010493.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171318.hg.1 |
TC1000010493.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000374401.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000536058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
ENST00000553795.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
NM_001004297.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171321.hg.1 |
TC1000010493.hg.1 |
Clariom D TC |