Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL832753.1 | - | 1 - 2 | 301 | 62 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | tubulin alpha 3c | TUBA3C |
TUBA2, bA408E5.3 |
55.5kb | Sense | Chr. 13 - 19173770 - 19181852 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens 1700 Probe spans exons |