Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB6 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY927631.1 | - | LIT3615 AY927631 |
1 - 2 | 299 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BPI fold containing family B member 2 | BPIFB2 |
BPIL1, C20orf184, LPLUNC2, RYSR, dJ726C3.2 |
-21.0 | Sense | Chr. 20 - 33007578 - 33023709 on NCBI Build GRCh38 |
Downstream | BPI fold containing family B member 3 | BPIFB3 |
C20orf185, LPLUNC3, RYA3 |
22.3kb | Sense | Chr. 20 - 33055331 - 33073637 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |