Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004060.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004060.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004060.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004056.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004056.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004056.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004056.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
ENST00000311042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
ENST00000383754.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
ENST00000447324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
NM_182935.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004048.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004061.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004061.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004061.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004055.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004055.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004055.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004055.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004059.hg.1 |
TC03000203.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004059.hg.1 |
ENST00000396228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004059.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004059.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149918.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000311042.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000383754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000447324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NM_001278322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NM_182935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000311042.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000383754.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000447324.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NM_001278322.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NM_182935.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
NR_103506.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149910.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149919.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149920.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149916.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000424090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000442631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000452959.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
ENST00000479860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_003090.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_103504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
NR_103505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149917.hg.1 |
TC0300007106.hg.1 |
Clariom D TC |