Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006397.hg.1 |
TC0X000487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006397.hg.1 |
ENST00000360035.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006397.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154275.hg.1 |
TC0X00007973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000360035.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
ENST00000416098.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007973.hg.1 |
NR_002216.1 |
Refseq |