Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF085855.1 | - | 1 | 447 | 113 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 474 | LINC00474 |
C9orf27, EST-YD1 |
-369.2kb | Anti-sense | Chr. 9 - 115888265 - 115925098 on NCBI Build GRCh38 |
Downstream | PAPPA antisense RNA 1 | PAPPA-AS1 |
DIPAS, NCRNA00156, PAPPA-AS, PAPPAAS, PAPPAS, TAF2C2, TAF4B, TAFII105 |
103.4kb | Anti-sense | Chr. 9 - 116398157 - 116400606 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |