Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000343999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000400023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000405854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000414287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000416411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000416411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000416411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000416411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000444628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000444628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000444628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000444628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000498829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000498829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000498829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
ENST00000498829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NM_001289922.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110522.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009107.hg.1 |
NR_110523.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009116.hg.1 |
ENST00000420902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009116.hg.1 |
ENST00000420902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009116.hg.1 |
ENST00000420902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009116.hg.1 |
ENST00000420902.1 |
Ensembl |