Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011544651.2 | XP_011542953.1 | 9 | 2285 | 84 | |
XM_011544652.2 | XP_011542954.1 | 10 | 2350 | 84 | ||
GenBank mRNA | AK026149.1 | - | 1 | 954 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sarcoglycan zeta | SGCZ | ZSG1 | -520.2kb | Anti-sense | Chr. 8 - 14084835 - 15238283 on NCBI Build GRCh38 |
Downstream | macrophage scavenger receptor 1 | MSR1 |
CD204, SCARA1, SR-A, SRA, phSR1, phSR2 |
347.2kb | Anti-sense | Chr. 8 - 16107878 - 16192791 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |