Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173533.hg.1 |
TC0700010345.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000399310.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000479493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010345.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
ENST00000407010.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
ENST00000609163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
NM_001101417.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
NM_001101426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700173534.hg.1 |
TC0700010345.hg.1 |
Clariom D TC |