Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42BPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK130237.1 | - | 1 | 747 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coenzyme Q8A | COQ8A |
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9 |
-72.5kb | Anti-sense | Chr. 1 - 226939339 - 226987545 on NCBI Build GRCh38 |
Downstream | basic transcription factor 3 pseudogene 9 | BTF3P9 | 372.7kb | Anti-sense | Chr. 1 - 227433913 - 227434606 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |