Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000056233.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000606261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
ENST00000607375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006977.hg.1 |
NM_004289.6 |
Refseq |