Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC027914.1 | - | 1 | 1252 | 110 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nectin cell adhesion molecule 3 | NECTIN3 |
CD113, CDW113, NECTIN-3, PPR3, PRR3, PVRL3, PVRR3 |
-272.9kb | Sense | Chr. 3 - 111071759 - 111201444 on NCBI Build GRCh38 |
Downstream | zinc finger BED-type containing 2 | ZBED2 | 48.9kb | Anti-sense | Chr. 3 - 111592900 - 111595335 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |