Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000468973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000468973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000482737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
ENST00000482737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000359511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000423538.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000468973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000468973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000469537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000482737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
ENST00000482737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_001286184.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NM_014655.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104408.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104411.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010159.hg.1 |
NR_104412.1 |
Refseq |