Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100505625 | LOC100505625 | -21.1kb | Anti-sense | Chr. 5 - 6686325 - 6707711 on NCBI Build GRCh38 | |
Downstream | microRNA 4278 | MIR4278 | 97.1kb | Anti-sense | Chr. 5 - 6827853 - 6827921 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |