Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000223951.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380110.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000380117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000397312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000423281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000440102.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000462649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
ENST00000544538.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_001171197.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009740.hg.1 |
NM_004432.4 |
Refseq |