Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000274341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000503117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000504713.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000508307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000510978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000514416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
ENST00000515590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011347.hg.1 |
NM_001884.3 |
Refseq |