Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4466 | MIR4466 | mir-4466 | -307.8kb | Anti-sense | Chr. 6 - 156779678 - 156779731 on NCBI Build GRCh38 |
Downstream | transmembrane protein 242 | TMEM242 |
BM033, C6orf35 |
201.2kb | Anti-sense | Chr. 6 - 157289022 - 157324259 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |