Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | brain derived neurotrophic factor | BDNF |
ANON2, BULN2 |
-34.4kb | Sense | Chr. 11 - 27654893 - 27722058 on NCBI Build GRCh38 |
Downstream |
heat shock protein 90 alpha family class A member 2, pseudogene |
HSP90AA2P |
HSP90AA2, HSP90ALPHA, HSPCA, HSPCAL3 |
131.4kb | Sense | Chr. 11 - 27888171 - 27891092 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |