Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4285 | MIR4285 | -10.6kb | Sense | Chr. 7 - 102293103 - 102293187 on NCBI Build GRCh38 | |
Downstream | speedy/RINGO cell cycle regulator family member E6 | SPDYE6 | 41.7kb | Anti-sense | Chr. 7 - 102345752 - 102356444 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |