Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin conjugating enzyme E2 Q2 | UBE2Q2 | -26.3kb | Sense | Chr. 15 - 75843286 - 75901047 on NCBI Build GRCh38 | |
Downstream | FBXO22 antisense RNA 1 | FBXO22-AS1 |
FBXO22OS, FISTC1OS, HsT18082, NCRNA00053 |
3.4kb | Anti-sense | Chr. 15 - 75932683 - 75933130 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |