Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, H/ACA box 11B |
SNORA11B | -38.4kb | Sense | Chr. 14 - 91126426 - 91126552 on NCBI Build GRCh38 | |
Downstream | G protein-coupled receptor 68 | GPR68 |
GPR12A, OGR1 |
65.4kb | Anti-sense | Chr. 14 - 91232532 - 91264581 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |