Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL713635.1 | - | 1 | 1951 | 99 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TNF alpha induced protein 8 | TNFAIP8 |
GG2-1, MDC-3.13, NDED, SCC-S2, SCCS2 |
-117.6kb | Sense | Chr. 5 - 119268692 - 119394604 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 170 member A | FAM170A | ZNFD | 114.4kb | Sense | Chr. 5 - 119629559 - 119635822 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |