Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sphingomyelin synthase 2 | SGMS2 | SMS2 | Sense | Chr. 4 - 107823645 - 107915047 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101929595 | LOC101929595 | 30.4kb | Anti-sense | Chr. 4 - 107863479 - 107932119 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |