Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | M-phase specific PLK1 interacting protein | MPLKIP |
ABHS, C7orf11, ORF20, TTD4 |
Anti-sense | Chr. 7 - 40132743 - 40134652 on NCBI Build GRCh38 | |
Downstream | succinyl-CoA:glutarate-CoA transferase | SUGCT |
C7orf10, DERP13, GA3, ORF19 |
357.0 | Sense | Chr. 7 - 40134887 - 40860767 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |