Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105373150 | LOC105373150 | Sense | Chr. X - 27153368 - 27399005 on NCBI Build GRCh38 | ||
Downstream |
protein phosphatase 4 regulatory subunit 3C, pseudogene |
PPP4R3CP |
FLFL3P, SMEK3P, smk1 |
65.2kb | Sense | Chr. X - 27460211 - 27463341 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |