Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 346 | LINC00346 |
C13orf29, NCRNA00346 |
-25.3kb | Sense | Chr. 13 - 110863987 - 110870308 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 431 | LINC00431 | 67.8kb | Anti-sense | Chr. 13 - 110966027 - 110990602 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |