Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1561 | LINC01561 | C10orf85 | -167.6kb | Anti-sense | Chr. 10 - 120597949 - 120600123 on NCBI Build GRCh38 |
Downstream | WD repeat domain 11 | WDR11 |
BRWD2, DR11, HH14, SRI1, WDR15 |
79.8kb | Anti-sense | Chr. 10 - 120851175 - 120909526 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |