Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MGC57346-CRHR1 readthrough | MGC57346-CRHR1 | Sense | Chr. 17 - 45620329 - 45835828 on NCBI Build GRCh38 | ||
Downstream | corticotropin releasing hormone receptor 1 | CRHR1 |
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L |
136.1kb | Sense | Chr. 17 - 45784280 - 45835828 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |