Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transient receptor potential cation channel subfamily V member 2 | TRPV2 |
VRL, VRL-1, VRL1 |
Anti-sense | Chr. 17 - 16415542 - 16437003 on NCBI Build GRCh38 | |
Downstream | LRRC75A antisense RNA 1 | LRRC75A-AS1 |
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19 |
22.3kb | Anti-sense | Chr. 17 - 16438987 - 16470648 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |