Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TAF1A antisense RNA 1 | TAF1A-AS1 | -66.2kb | Anti-sense | Chr. 1 - 222589920 - 222592633 on NCBI Build GRCh38 | |
Downstream |
axin interactor, dorsalization associated |
AIDA | C1orf80 | 6.5kb | Sense | Chr. 1 - 222668013 - 222712522 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |