Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | golgi to ER traffic protein 4 | GET4 |
C7orf20, CEE, CGI-20, TRC35 |
Anti-sense | Chr. 7 - 876554 - 896434 on NCBI Build GRCh38 | |
Downstream | ArfGAP with dual PH domains 1 | ADAP1 |
CENTA1, GCS1L, p42IP4 |
11.4kb | Sense | Chr. 7 - 897900 - 955407 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |