Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proteasome assembly chaperone 2 | PSMG2 |
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1 |
Sense | Chr. 18 - 12658739 - 12725740 on NCBI Build GRCh38 | |
Downstream | centrosomal protein 76 | CEP76 |
C18orf9, HsT1705 |
1.3kb | Anti-sense | Chr. 18 - 12661733 - 12702777 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |