Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cadherin like and PC-esterase domain containing 1 | CPED1 | C7orf58 | Sense | Chr. 7 - 120988697 - 121297444 on NCBI Build GRCh38 | |
Downstream | Wnt family member 16 | WNT16 | 30.6kb | Sense | Chr. 7 - 121325367 - 121341104 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |