Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GRB2 associated regulator of MAPK1 subtype 1 | GAREM1 |
C18orf11, FAM59A, GAREM, Gm944 |
Sense | Chr. 18 - 32263521 - 32470518 on NCBI Build GRCh38 | |
Downstream | WW domain binding protein 11 pseudogene 1 | WBP11P1 | HsT3017 | 198.7kb | Anti-sense | Chr. 18 - 32511663 - 32514634 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |