Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bromodomain adjacent to zinc finger domain 1A | BAZ1A |
ACF1, WALp1, WCRF180, hACF1 |
Sense | Chr. 14 - 34752731 - 34875647 on NCBI Build GRCh38 | |
Downstream | immunoglobulin (CD79A) binding protein 1 pseudogene 1 | IGBP1P1 | C14orf19 | 175.7kb | Anti-sense | Chr. 14 - 34939922 - 34940496 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |