Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HEAT repeat containing 9 | HEATR9 | C17orf66 | Anti-sense | Chr. 17 - 35854946 - 35868891 on NCBI Build GRCh38 | |
Downstream | C-C motif chemokine ligand 5 | CCL5 |
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP |
6.9kb | Anti-sense | Chr. 17 - 35871491 - 35880373 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |