Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | receptor-like tyrosine kinase | RYK |
D3S3195, JTK5, JTK5A, RYK1 |
-96.4kb | Sense | Chr. 3 - 134157133 - 134250834 on NCBI Build GRCh38 |
Downstream | ribosomal protein L39 pseudogene 5 | RPL39P5 | RPL39_7_406 | 2.5kb | Sense | Chr. 3 - 134351739 - 134352143 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |