Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011609.hg.1 |
ENST00000458184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011609.hg.1 |
ENST00000458184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366934.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000366935.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000470014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000485274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
ENST00000489246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_001297754.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017321.hg.1 |
NM_018040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011609.hg.1 |
ENST00000458184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011609.hg.1 |
ENST00000458184.1 |
Ensembl |