Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DNA replication and sister chromatid cohesion 1 | DSCC1 | DCC1 | -16.9kb | Sense | Chr. 8 - 119833941 - 119855930 on NCBI Build GRCh38 |
Downstream | collagen type XIV alpha 1 chain | COL14A1 | UND | 250.6kb | Anti-sense | Chr. 8 - 120124468 - 120373575 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |