Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, H/ACA box 72 |
SNORA72 |
RNU72, U72 |
Sense | Chr. 8 - 98042086 - 98042217 on NCBI Build GRCh38 | |
Downstream | glutamate rich 5 | ERICH5 | C8orf47 | 21.2kb | Anti-sense | Chr. 8 - 98064522 - 98093610 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |