Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005278385.4 | XP_005278442.1 | 6 | 2504 | 112 | |
GenBank mRNA | BC034279.1 | - | 1 | 653 | 112 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carcinoembryonic antigen related cell adhesion molecule 3 | CEACAM3 |
CD66D, CEA, CGM1, W264, W282 |
-22.1kb | Anti-sense | Chr. 19 - 41796590 - 41811553 on NCBI Build GRCh38 |
Downstream | DMRT like family C2 | DMRTC2 | 8.8kb | Anti-sense | Chr. 19 - 41844743 - 41852330 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |