Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
240115_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000458178.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006743.hg.1 |
ENST00000538634.1 |
Ensembl |
Human Genome U95 Set |
46430_at |
- |
- |
Human Genome U95 Set |
46430_PM_at |
- |
- |
Human Genome U133 Set |
240115_at |
- |
- |
Human Genome U133 Set |
240115_PM_at |
- |
- |