Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR933684.1 | - | 3 | 2755 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 6 | CCDC6 |
D10S170, H4, PTC, TPC, TST1 |
-48.8kb | Sense | Chr. 10 - 59788747 - 59906656 on NCBI Build GRCh38 |
Downstream |
ankyrin 3, node of Ranvier (ankyrin G) |
ANK3 |
ANKYRIN-G, MRT37 |
67.0kb | Sense | Chr. 10 - 60026298 - 60733526 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |